Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC129 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554978
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554978 Supplier Invitrogen™ Supplier No. PA554978
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LFSRDMSEEE REEAEQLQTL REALRQQVAE LEFQLGDRAQ QIREGILLQL EVLTAEPPEH YSNLHQYNWI EESNGQTSCS KIHPGMAPRT VFPPDDGQEA PCSGGTQLAA FTPPTLENST RMSPSSSAWA KLGPTPLSNC PVGEKDAD Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 54%.

CCDC129 gene ontology annotations related to this gene include signaling receptor binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC129
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC129
Gene Accession No. Q6ZRS4
Gene Alias CCDC129; coiled-coil domain containing 129; coiled-coil domain-containing protein 129; D530004J12Rik; Gm458; Itprid1; ITPR-interacting domain-containing protein 1; Protein ITPRID1; RGD1561131
Gene Symbols CCDC129
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC129. Recombinant protein control fragment (Product #RP-92814).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223075
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.