Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC129 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988281
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8281 Supplier Bioss Supplier No. BS8107R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC129 gene ontology annotations related to this gene include signaling receptor binding.

Specifications

Antigen CCDC129
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCDC129
Gene Accession No. Q6ZRS4
Gene Alias CCDC129; coiled-coil domain containing 129; Coiled-coil domain-containing protein 129; ITPRID1; ITPR-interacting domain-containing protein 1; Protein ITPRID1
Gene Symbols CCDC129
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC129.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223075
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.