Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC132 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118355
This item is not returnable.
View return policy
Description
CCDC132 Polyclonal Antibody for Western Blot, IHC (P), IP
CCDC132 is a protein coding gene.
Specifications
| CCDC132 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q96JG6 | |
| Vps50 | |
| E. coli-derived Human VPS50/CCDC132 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Vps50 | |
| 1700034M03Rik; 8430415E05Rik; Bcl2-like protein blm; BLM; C87205; CCDC132; coiled-coil domain containing 132; coiled-coil domain-containing protein 132; DKFZp313I2429; EARP/GARPII complex subunit VPS50; FLJ20097; KIAA1861; mKIAA1861; RGD1304751; syndetin; Vps50; VPS50 EARP/GARPII complex subunit; VPS50, EARP/GARPII complex subunit; VPS54L | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 55610 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction