Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC132 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501564847
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4847 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4847 Supplier Bethyl Laboratories, Inc Supplier No. A303324AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Chicken

CCDC132 is a protein coding gene.

Specifications

Antigen CCDC132
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene VPS50
Gene Accession No. Q96JG6
Gene Alias CCDC132, coiled-coil domain containing 132, coiled-coil domain-containing protein 132, DKFZp313I2429, EARP/GARPII complex subunit VPS50, FLJ20097, KIAA1861, syndetin, VPS50 EARP/GARPII complex subunit, VPS54L
Gene Symbols VPS50
Host Species Rabbit
Immunogen Between 914 and 964
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55610
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.