Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC134 Antibody (04), Janelia Fluor™ 549 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306134JF549
Description
CCDC134 Monoclonal antibody specifically detects CCDC134 in Human samples. It is validated for ELISASpecifications
CCDC134 | |
Monoclonal | |
Janelia Fluor 549 | |
coiled-coil domain containing 134, coiled-coil domain-containing protein 134, dJ821D11.3, FLJ22349, MGC21013 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCDC134 (Q9H6E4; Met 1-Leu 229). | |
0.1 mL | |
Cancer | |
79879 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
04 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction