Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC135 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570881 Supplier Invitrogen™ Supplier No. PA570881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Component of the nexin-dynein regulatory complex (N-DRC) a key regulator of ciliary/flagellar motility which maintains the alignment and integrity of the distal axoneme and regulates microtubule sliding in motile axonemes. Involved in the regulation of flagellar motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC135
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DRC7
Gene Accession No. Q8IY82
Gene Alias C16orf50; CCDC135; CFAP50; coiled-coil domain containing 135; coiled-coil domain-containing protein 135; Coiled-coil domain-containing protein lobo homolog; DRC7; Dynein regulatory complex subunit 7; FAP50; Gm770; RGD1565601; spermatogenesis-related gene in late stages of spermatogenesis cells protein; Srgl; SRG-L; SRG-S; Unknown (protein for MGC:134162)
Gene Symbols DRC7
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human CCDC135.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84229
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.