Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC136 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988268
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8268 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8268 Supplier Bioss Supplier No. BS8090R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in acrosome formation in spermatogenesis and in fertilization.

Specifications

Antigen CCDC136
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCDC136
Gene Accession No. Q96JN2
Gene Alias CCDC136; coiled-coil domain containing 136; Coiled-coil domain-containing protein 136; KIAA1793; NAG6; Nasopharyngeal carcinoma-associated gene 6 protein
Gene Symbols CCDC136
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC136/NAG6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64753
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.