Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC137 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116781 Supplier Thermo Scientific Supplier No. PA5116781
Add to Cart
Add to Cart

CCDC137 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC137
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CCDC137
Gene Accession No. Q6PK04
Gene Alias CCDC137; coiled-coil domain containing 137; coiled-coil domain-containing protein 137; cPERP-B; hepatocellular carcinoma related protein 2; RaRF; retinoic acid resistance factor
Gene Symbols CCDC137
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 213-288 of human CCDC137 (NP_9549811).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339230
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.