Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC138 Polyclonal Antibody

Catalog No. PIPA562083
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562083 Supplier Invitrogen™ Supplier No. PA562083

Rabbit Polyclonal Antibody

Immunogen sequence: IKGVEEEVLT RFQIIKEQHD AEVEHLTEVL KEKNKETKRL RSSFDALKEL NDTLKKQLNE ASEENRKIDI QAKRVQARLD NLQRKYEFMT IQRLKGSSHA VH Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

The function of the CCDC138 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC138
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC138
Gene Accession No. Q96M89
Gene Alias 6230424H07Rik; BC042726; Ccdc138; coiled-coil domain containing 138; coiled-coil domain-containing protein 138; RGD1566050
Gene Symbols CCDC138
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC138. Recombinant protein control fragment (Product #RP-99681).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165055
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.