Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112950
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112950 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112950 Supplier Invitrogen™ Supplier No. PA5112950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC14 Polyclonal Antibody for Western Blot, IHC (P), ELISA

CCDC14 (Coiled-Coil Domain-Containing 14) is a gene that encodes a protein involved in ciliary function and cellular division processes. This protein contains coiled-coil domains that facilitate protein-protein interactions crucial for the assembly and stabilization of microtubule structures within the cell. Research indicates that CCDC14 is essential for proper ciliogenesis, the process by which cells form cilia, which are vital for cell motility, fluid movement, and signal transduction. Additionally, studies have revealed its significant role in the regulation of cell cycle progression and cytokinesis. CCDC14 is required for the accurate segregation of chromosomes during cell division, ensuring genomic stability. Mutations or dysregulation of CCDC14 can lead to ciliopathies and other cellular dysfunctions, highlighting its critical role in maintaining normal cellular activities and its potential implications in various diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC14
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CCDC14
Gene Accession No. Q49A88
Gene Alias CCDC14; coiled-coil domain containing 14; coiled-coil domain-containing protein 14
Gene Symbols CCDC14
Host Species Rabbit
Immunogen Recombinant Human Coiled-coil domain-containing protein 14 protein (159-440AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64770
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.