Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC141 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116237
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116237 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116237 Supplier Invitrogen™ Supplier No. PA5116237
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC141 Polyclonal Antibody for Western Blot

Coiled-coil domain-containing protein 141 (CCDC141), also known as CAMDI, is a recently identified disrupted in schizophrenia 1 (DISC1)-binding protein that also associates with myosin II. CCDC141 preferentially associates with phosphomyosin II and causes an accumulation of phosphomyosin II at the centrosome in a DISC1-dependent manner. Knockdown of CCDC141 expression by RNAi led to severely impaired radial migration with disoriented chromosomes, suggesting that CCDC141 is required for radial migration through DISC1 and myosin II-mediated centrosome positioning during neuronal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC141
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CCDC141
Gene Accession No. Q6ZP82
Gene Alias 2610301F02Rik; CAMDI; CCDC141; coiled-coil domain containing 141; coiled-coil domain-containing protein 141; Coiled-coil protein associated with myosin II and DISC1; ENSMUSG00000075261
Gene Symbols CCDC141
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human CCDC141.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285025
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.