Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC144A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565466
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565466 Supplier Invitrogen™ Supplier No. PA565466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ACPEEEPLLDN STRGTDVKDI PFNLTNNIPG CEEEDASEIS VSVVFETFPE QKEPSLKNII HPYYHPYSGS QEHVCQSSSK Highest antigen sequence identity to the following orthologs - mouse 27%, rat 27%.

CCDC144A (Coiled-Coil Domain Containing 144A) is a Protein Coding gene. An important paralog of this gene isENSG00000266302.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC144A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC144A
Gene Accession No. A2RUR9
Gene Alias CCDC144A; coiled-coil domain containing 144A; coiled-coil domain-containing protein 144A; KIAA0565
Gene Symbols CCDC144A
Host Species Rabbit
Immunogen Recombinant Human CCDC144A. Recombinant protein control fragment (Product #RP-105614).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9720
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.