Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC147 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140138 Supplier Invitrogen™ Supplier No. PA5140138
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: KEEIVNLTKL VEQGSGLSMD QHSNIRDLLR FKEEVTKERD QLLSEVVKLR ESLAQTTEQQ QETERSKEEA EHAISQFQQE IQQRQN Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 87%.

CFAP58 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC147
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP58
Gene Accession No. Q5T655
Gene Alias bA127L20.4; bA127L20.4 (novel protein); bA127L20.5; bA127L20.5 (novel protein); bA554P13.1; C10orf80; CCDC147; Cfap58; cilia and flagella associated protein 58; cilia- and flagella-associated protein 58; coiled-coil domain containing 147; coiled-coil domain-containing protein 147; coiled-coil domain-containing protein 147-like; Gm969
Gene Symbols CFAP58
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CFAP58. Recombinant protein control fragment (Product #RP-109271).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 159686
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.