Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC147 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988288
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8288 Supplier Bioss Supplier No. BS8118R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CFAP58 is a protein coding gene.

Specifications

Antigen CCDC147
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CFAP58
Gene Accession No. Q5T655
Gene Alias bA127L20.4; bA127L20.4 (novel protein); bA127L20.5; bA127L20.5 (novel protein); bA554P13.1; C10orf80; CCDC147; CFAP58; cilia and flagella associated protein 58; Cilia- and flagella-associated protein 58; coiled-coil domain containing 147; Coiled-coil domain-containing protein 147
Gene Symbols CFAP58
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC147.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 159686
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.