Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC153 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559594
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559594 Supplier Invitrogen™ Supplier No. PA559594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: REEAEQALGE RDQALAQLRA HMADMEAKYE EILHDSLDRL LAKLRAIKQQ WDGAALRLHA RHKEQQRQFG LTPPGSLRPP APSL Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 63%.

CCDC153's properties have yet to be elucidated. However, research suggests that it may be a neuronal subtype marker. It has a molecular weight of 24 kDa and is often observed on a Western blot as a dimer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC153
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC153
Gene Accession No. Q494R4
Gene Alias Ccdc153; coiled-coil domain containing 153; coiled-coil domain-containing protein 153; RGD1305351
Gene Symbols CCDC153
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC153. Recombinant protein control fragment (Product #RP-97545).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283152
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.