Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC153 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988290
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8290 Supplier Bioss Supplier No. BS8121R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC153′s properties have yet to be elucidated. However, research suggests that it may be a neuronal subtype marker. It has a molecular weight of 24 kDa and is often observed on a Western blot as a dimer.

Specifications

Antigen CCDC153
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCDC153
Gene Accession No. Q494R4
Gene Alias CCDC153; coiled-coil domain containing 153; Coiled-coil domain-containing protein 153; RGD1305351
Gene Symbols CCDC153
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Human CCDC153.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283152
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.