Learn More
Invitrogen™ CCDC158 Polyclonal Antibody

Description
CCDC158 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | CCDC158 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Ccdc158 |
| Gene Accession No. | Q5M9N0, Q8CDI6 |
| Gene Alias | 4932413O14Rik; Ccdc158; coiled-coil domain containing 158; coiled-coil domain-containing protein 158; RGD1308523 |
| Gene Symbols | Ccdc158 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.