Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC159 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565888 Supplier Invitrogen™ Supplier No. PA565888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQGEKEEHKWG MEQGRQELYG ALTQGLQGLE KTLRDSEEMQ RARTTRCLQL LAQEIRDSKK FLWEELELVR EEVTFIYQKL QA Highest antigen sequence identity to the following orthologs - mouse 81%, rat 80%.

CCDC159 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC159
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC159
Gene Accession No. P0C7I6
Gene Alias CCDC159; coiled-coil domain containing 159; coiled-coil domain-containing protein 159
Gene Symbols CCDC159
Host Species Rabbit
Immunogen Recombinant Human CCDC159. Recombinant protein control fragment (Product #RP-106561).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126075
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.