Learn More
CCDC16 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301418A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Located on chromosome 17, this gene encodes for a coiled-coil domain containing 16 protein with unknown function.Specifications
| CCDC16 | |
| Polyclonal | |
| Unconjugated | |
| ZNF830 | |
| CCDC16, coiled-coil domain containing 16, coiled-coil domain-containing protein 16, hypothetical protein MGC20398, MGC20398, OMCG1, orphan maintece of genome 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91603 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q96NB3 | |
| ZNF830 | |
| Between 150 and 200 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.