Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC160 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560793
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560793 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560793 Supplier Invitrogen™ Supplier No. PA560793
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSSRKFQEES KFKRKKYIFQ LNEIEQEQNL RENKRNISKN ETDTNSASYE SSNVDVTTEE SFNSTEDNST CSTDNLPALL RQDIRKKFME RM Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 46%.

CCDC160 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC160
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC160
Gene Accession No. A6NGH7
Gene Alias CCDC160; coiled-coil domain containing 160; coiled-coil domain-containing protein 160; UPF0625 coiled-coil domain-containing protein ENSP00000359845
Gene Symbols CCDC160
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC160. Recombinant protein control fragment (Product #RP-100311).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347475
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.