Learn More
Invitrogen™ CCDC168 Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RNATGSAVSC ETQISEDFVD IQTDIESPAD LDECSCLEVS ESEECVFLEA NSYLSQESEN ILFELQTGIP LENVYKITTD LKSFYSEDSG SHCT Highest antigen sequence indentity to the following orthologs: Mouse - 40%, Rat - 31%.
Specifications
Specifications
| Antigen | CCDC168 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CCDC168 |
| Gene Accession No. | Q8NDH2 |
| Gene Alias | C13orf40; CCDC168; coiled-coil domain containing 168; coiled-coil domain-containing protein 168 |
| Gene Symbols | CCDC168 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.