Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531704 Supplier Invitrogen™ Supplier No. PA531704
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: MCF-7. Predicted reactivity: Human (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CCDC17 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene CCDC17
Gene Accession No. Q96LX7
Gene Alias Ccdc17; coiled-coil domain containing 17; coiled-coil domain-containing protein 17
Gene Symbols CCDC17
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CCDC17. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149483
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.