Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC170 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23786025UL
Description
CCDC170 Polyclonal antibody specifically detects CCDC170 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
CCDC170 | |
Polyclonal | |
Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
Q8IYT3 | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
80129 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol | |
bA282P11.1, C6orf97, chromosome 6 open reading frame 97, coiled-coil domain containing 170, FLJ23305, hypothetical protein LOC80129 | |
This antibody was developed against a recombinant protein corresponding to amino acids: RMVSQLEAQISELVEQLGKESGFHQKALQRAQKAENMLETLQGQLTHLEAELVSGGVLRDNLNFEKQKYLKFLDQLSQKMKLDQMA | |
25 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction