Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC171 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554084
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554084 Supplier Invitrogen™ Supplier No. PA554084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KAQAAQSESE LQKLSQAFHK DAEEKLTFLH TLYQHLVAGC VLIKQPEGML DKFSWSELCA VLQENVDALI ADLNRANEKI RHLEYICKNK SDTMREL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 26%.

Chromosome 9 consists of about 145 million bases and 4% of the human genome and encodes nearly 900 genes. Considered to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterized by harmful vascular defects, is associated with the chromosome 9 gene encoding endoglin protein, ENG. Familial dysautonomia is also associated with chromosome 9 though through the gene IKBKAP. Notably, chromosome 9 encompasses the largest interferon family gene cluster. Chromosome 9 is partnered with chromosome 22 in the translocation leading to the aberrant production of BCR-ABL fusion protein often found in leukemias. There are two isoforms of C9orf93 that are produced as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC171
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC171
Gene Accession No. Q6TFL3
Gene Alias bA536D16.1; bA778P13.1; C9orf93; CCDC171; coiled-coil domain containing 171; coiled-coil domain-containing protein 171; myosin tail domain containing protein
Gene Symbols CCDC171
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC171. Recombinant protein control fragment (Product #RP-93030).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203238
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.