Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC173 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | C2orf77 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CCDC173 Polyclonal specifically detects CCDC173 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
C2orf77 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CCDC173, Chromosome 2 Open Reading Frame 77, Coiled-Coil Domain Containing 173, Coiled-Coil Domain-Containing Protein 173, Hypothetical LOC129881 | |
CCDC173 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
Q0VFZ6 | |
129881 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MLVRFGRRCGRAKESTEIRNSEEDQVLYLPLLPSKVDLQQVTIIPHDEWKRIQDSLDRLTREAACLRAERKAKKEMHLRSQEVVKHWTNTYAGMKEQTLEAK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title