Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC173 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $728.30
Specifications
| Antigen | C2orf77 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CCDC173 Polyclonal specifically detects CCDC173 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| C2orf77 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CCDC173, Chromosome 2 Open Reading Frame 77, Coiled-Coil Domain Containing 173, Coiled-Coil Domain-Containing Protein 173, Hypothetical LOC129881 | |
| CCDC173 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q0VFZ6 | |
| 129881 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TIAEYRAIVMKNKEEEERQRKIEAKEQLLAVMKADQIFWEHEKEKKCKADKEHQEVQDAHIQQMAKNKFNAKQAKQAELDYCRLT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title