Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC174 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570256
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Probably involved in neuronal development.
Specifications
CCDC174 | |
Polyclonal | |
Unconjugated | |
CCDC174 | |
C130022K22Rik; C20H3orf19; C22H3orf19; C3orf19; Ccdc174; coiled-coil domain containing 174; coiled-coil domain-containing protein 174; HSPC212; IHPM; IHPMR; RGD1305225; Unknown (protein for MGC:139858) | |
Rabbit | |
Affinity chromatography | |
RUO | |
297458 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5PQS7 | |
CCDC174 | |
synthetic peptide directed towards the middle region of rat CCDC174. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction