Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC176 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551822
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551822 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551822 Supplier Invitrogen™ Supplier No. PA551822
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGEAKMPSKG KDKKKGKSKG KDTKKLIKTD ESVVDRAKAN ASLWEARLEV TELSRIKYRD TSRILAKSNE DLKKKQCKME KDIMSVLSYL KKQDQEKDNM Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 68%.

Basal body protein required in multiciliate cells to align and maintain cilia orientation in response to flow. May act by mediating a maturation step that stabilizes and aligns cilia orientation. Not required to respond to planar cell polarity (PCP) or flow-based orientation cues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC176
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BBOF1
Gene Accession No. Q8ND07
Gene Alias 2900006K08Rik; AI427784; basal body orientation factor 1; basal body-orientation factor 1; BBOF1; C10H14orf45; C14orf45; C8H14orf45; Ccdc176; coiled-coil domain containing 176; coiled-coil domain-containing protein 176; RGD1560978; uncharacterized protein C14orf45 homolog
Gene Symbols BBOF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC176. Recombinant protein control fragment (Product #RP-89099).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80127
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.