Learn More
Invitrogen™ CCDC176 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551833
Description
Immunogen sequence: ISRKHYKQIA QAAFNLKMRA ACTGRTEYPK IRTFDGREHS TNSVNQDLLE AEKWTHIEGN VDIGDLTWEQ KEKVLRLLFA KMNGCPSRKY NQSSRPPVPD YVVSDSGETK EFGDESKLQD KIFITQQIAI SDSSGEVVLP TIPKEPQ Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.
Basal body protein required in multiciliate cells to align and maintain cilia orientation in response to flow. May act by mediating a maturation step that stabilizes and aligns cilia orientation. Not required to respond to planar cell polarity (PCP) or flow-based orientation cues.
Specifications
CCDC176 | |
Polyclonal | |
Unconjugated | |
BBOF1 | |
2900006K08Rik; AI427784; basal body orientation factor 1; basal body-orientation factor 1; BBOF1; C10H14orf45; C14orf45; C8H14orf45; Ccdc176; coiled-coil domain containing 176; coiled-coil domain-containing protein 176; RGD1560978; uncharacterized protein C14orf45 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80127 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8ND07 | |
BBOF1 | |
Recombinant protein corresponding to Human CCDC176. Recombinant protein control fragment (Product #RP-89083). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.