Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC18 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178738V
Description
CCDC18 Polyclonal antibody specifically detects CCDC18 in Human samples. It is validated for Western BlotSpecifications
CCDC18 | |
Polyclonal | |
Western Blot | |
coiled-coil domain containing 18, coiled-coil domain-containing protein 18, dJ717I23.1, NY-SAR-41, sarcoma antigen NY-SAR-41 | |
The immunogen for this product maps to a region between residue 225 and 275 of human Coiled-Coil Domain Containing 18 using the numbering given in entry CAI22508.1 (GeneID 343099). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
343099 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction