Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC18 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988350
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8350 Supplier Bioss Supplier No. BS8189R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Coiled-coil domain-containing protein 18 (CCDC18) is an uncharacterized protein identified as an antigen recognized by serum antibodies of a sarcoma cancer immunome.

Specifications

Antigen CCDC18
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCDC18
Gene Accession No. Q5T9S5
Gene Alias CCDC18; coiled-coil domain containing 18; Coiled-coil domain-containing protein 18; NY-SAR-41; Sarcoma antigen NY-SAR-24; sarcoma antigen NY-SAR-41
Gene Symbols CCDC18
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC18.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343099
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.