Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC184 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559003
Description
Immunogen sequence: MEDGLLEIMT KDGGDMPAPL EVSTVPAVGD VISGEYNGGM KELMEHLKAQ LQ Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
CCDC184 is a protein coding gene.
Specifications
| CCDC184 | |
| Polyclonal | |
| Unconjugated | |
| CCDC184 | |
| 5630400A08; AI836003; C12orf68; C27H12orf68; C5H12orf68; C6H12orf68; Ccdc184; coiled-coil domain containing 184; coiled-coil domain-containing protein 184; uncharacterized protein C12orf68 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 387856 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q52MB2 | |
| CCDC184 | |
| Recombinant protein corresponding to Human CCDC184. Recombinant protein control fragment (Product #RP-97855). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction