Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC185 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569908
Description
This target displays homology in the following species: Human: 100%.
CCDC185 is a protein coding gene.
Specifications
CCDC185 | |
Polyclonal | |
Unconjugated | |
CCDC185 | |
C1orf65; CCDC185; coiled-coil domain containing 185; coiled-coil domain-containing protein 185 | |
Rabbit | |
Affinity chromatography | |
RUO | |
164127 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N715 | |
CCDC185 | |
synthetic peptide directed towards the N terminal region of human CCDC185. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction