Learn More
CCDC186 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305727AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Drosophila Deformed epidermal autoregulatory factor 1 (DEAF-1) is a DNA-binding protein that interacts with regulatory sequences first described in the Deformed epidermal autoregulatory element.Specifications
CCDC186 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q7Z3E2 | |
CCDC186 | |
Between 625 and 675 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
CCDC186 | |
C10orf118, chromosome 10 open reading frame 118, CTCL tumor antigen HD-CL-01/L14-2, CTCL tumor antigen L14-2, FLJ10188, FLJ35301 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55088 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.