Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC187 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570173
Description
This target displays homology in the following species: Cow: 92%; Dog: 92%; Horse: 100%; Human: 100%; Rat: 92%.
CCDC187 is a protein coding gene.
Specifications
CCDC187 | |
Polyclonal | |
Unconjugated | |
CCDC187 | |
CCDC187; coiled-coil domain containing 187; coiled-coil domain-containing protein 187; RGD1565512; similar to hypothetical protein 4932418E24; uncharacterized MGC50722; uncharacterized protein LOC499759 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
399693 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A0A096LP49 | |
CCDC187 | |
synthetic peptide directed towards the N terminal region of human CCDC187. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction