Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | CCDC19 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CCDC19 Polyclonal specifically detects CCDC19 in Human samples. It is validated for Western Blot.Specifications
CCDC19 | |
Polyclonal | |
Rabbit | |
Q5VU18 | |
25790 | |
Synthetic peptides corresponding to CCDC19(coiled-coil domain containing 19) The peptide sequence was selected from the N terminal of CCDC19. Peptide sequence MPLSTAGILSSSSAASNRSRNKARYRTKAVSSEVDESLFGDIKSPAQGQS. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
coiled-coil domain containing 19, coiled-coil domain-containing protein 19, mitochondrial, nasopharyngeal epithelium specific protein 1 (NESG1), NESG1Nasopharyngeal epithelium-specific protein 1 | |
CCDC19 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title