Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543685
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543685 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543685 Supplier Invitrogen™ Supplier No. PA543685
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MPLSTAGILS SSSAASNRSR NKARYRTKAV SSEVDESLFG DIKSPAQGQS Sequence homology: Cow: 85%; Dog: 85%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Rat: 85%; Yeast: 79%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC19
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CFAP45
Gene Accession No. Q9UL16
Gene Alias 1700028D05Rik; CCDC19; CFAP45; cilia and flagella associated protein 45; cilia- and flagella-associated protein 45; coiled-coil domain containing 19; Coiled-coil domain-containing protein 19; coiled-coil domain-containing protein 19, mitochondrial; nasopharyngeal epithelium specific protein 1; nasopharyngeal epithelium specific protein 1 (NESG1); nasopharyngeal epithelium-specific protein 1; NESG1; protein CFAP45, mitochondrial; Unknown (protein for MGC:134208)
Gene Symbols CFAP45
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CCDC19 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25790
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.