Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560439
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560439 Supplier Invitrogen™ Supplier No. PA560439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEATMDAVMT RKKIMKQKEM VWNNNKKLSD LEEVAKERAQ NLLQRANKLR MEQEEELKDM SKIILNAKCH AIRDAQILEK QQI Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC19
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP45
Gene Accession No. Q9UL16
Gene Alias 1700028D05Rik; CCDC19; CFAP45; cilia and flagella associated protein 45; cilia- and flagella-associated protein 45; coiled-coil domain containing 19; Coiled-coil domain-containing protein 19; coiled-coil domain-containing protein 19, mitochondrial; nasopharyngeal epithelium specific protein 1; nasopharyngeal epithelium specific protein 1 (NESG1); nasopharyngeal epithelium-specific protein 1; NESG1; protein CFAP45, mitochondrial; Unknown (protein for MGC:134208)
Gene Symbols CFAP45
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC19. Recombinant protein control fragment (Product #RP-99841).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25790
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.