Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC21 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178219V
Description
CCDC21 Polyclonal antibody specifically detects CCDC21 in Human samples. It is validated for Western BlotSpecifications
CCDC21 | |
Polyclonal | |
Western Blot | |
coiled-coil domain containing 21, coiled-coil domain-containing protein 21, DKFZp434L0117, DKFZp434P232, FLJ13976, FLJ22000 | |
The immunogen for this product maps to a region between residue 675 and 725 of human Centrosomal Protein 85kDa using the numbering given in entry NP_073615.2 (GeneID 64793). | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication | |
64793 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction