Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555806
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555806 Supplier Invitrogen™ Supplier No. PA555806
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLLEETQAIC REKEIQLESL RQREAEFSSA GHSLQDKQSV EETSGEGPEV EMESWQKRYD SLQKIVEKQQ QKMDQLRSQV QSLEQEVAQE EGTSQ Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.

This gene encodes a protein that belongs to the centrosome-associated family of proteins. The centrosome is a subcellular organelle in the animal cell that functions as a microtubule organizing center and is involved in cell-cycle progression. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC21
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP85
Gene Accession No. Q6P2H3
Gene Alias 2410030J07Rik; AI173272; AI173504; CCDC21; centrosomal protein 85; centrosomal protein 85kDa; centrosomal protein of 85 kDa; CEP85; coiled-coil domain containing 21; coiled-coil domain-containing protein 21; RGD1311520
Gene Symbols CEP85
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC21. Recombinant protein control fragment (Product #RP-94183).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64793
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.