Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC22 Monoclonal Antibody (OTI2E4)
GREENER_CHOICE

Catalog No. PIMA527398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527398 Supplier Invitrogen™ Supplier No. MA527398
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCDC22 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

CCDC22 is a recently identified coiled-coil domain-containing protein that has been shown to bind copines, which are calcium-dependent, membrane-binding proteins that may function in calcium signaling. In rat, CCDC22 has been observed to localize in multiple regions of the brain, including the prefrontal and somatosensory cortex, dentate gyrus and thalamus, and in the ipsilateral motor neurons of the spinal cord after sciatic nerve transection suggesting that it may play a role in neuronal injury response. The human CCDC22 gene has been identified as a novel candidate gene for syndromic X-linked intellectual disability (XLID).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC22
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ccdc22
Gene Accession No. O60826
Gene Alias AI481216; Ccdc22; ccdc22 {ECO:0000250; coiled-coil domain containing 22; coiled-coil domain-containing protein 22; coiled-coil domain-containing protein 22 {ECO:0000250; CXorf37; DXImx40e; JM1; Ppp1r3f; RGD1560910; RTSC2; Sfc22; UniProtKB:O60826}
Gene Symbols Ccdc22
Host Species Mouse
Immunogen Full length human recombinant protein of CCDC22 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28952
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.