Learn More
CCDC22 Mouse anti-Human, Clone: OTI2A9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811120

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CCDC22 is a recently identified coiled-coil domain-containing protein that has been shown to bind copines, which are calcium-dependent, membrane-binding proteins that may function in calcium signaling. In rat, CCDC22 has been observed to localize in multiple regions of the brain, including the prefrontal and somatosensory cortex, dentate gyrus and thalamus, and in the ipsilateral motor neurons of the spinal cord after sciatic nerve transection suggesting that it may play a role in neuronal injury response. The human CCDC22 gene has been identified as a novel candidate gene for syndromic X-linked intellectual disability (XLID).Specifications
CCDC22 | |
Monoclonal | |
Unconjugated | |
CCDC22 | |
CXorf37, JM1, RTSC2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
28952 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2A9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O60826 | |
CCDC22 | |
Full length human recombit protein of human CCDC22 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.