Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552569
Description
Immunogen sequence: DPPARKEKTK VKESVSRVEK AKQKSAQQEL KQRQRAEIYA LNRVMTELEQ QQFDEFCKQM QPPGE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Specifications
CCDC23 | |
Polyclonal | |
Unconjugated | |
SVBP | |
2410005K17Rik; AI851162; Ccdc23; coiled coil domain-containing protein 23; coiled-coil domain containing 23; coiled-coil domain-containing protein 23; RGD1311232; small vasohibin binding protein; small vasohibin-binding protein; SVBP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
374969 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N300 | |
SVBP | |
Recombinant protein corresponding to Human CCDC23. Recombinant protein control fragment (Product #RP-89736). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction