Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113183
Description
CCDC24 Polyclonal Antibody for Western Blot, IHC (P), ELISA
CCDC24 is a protein coding gene.
Specifications
| CCDC24 | |
| Polyclonal | |
| Unconjugated | |
| CCDC24 | |
| CCDC24; coiled-coil domain containing 24; coiled-coil domain-containing protein 24 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 149473 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8N4L8 | |
| CCDC24 | |
| Recombinant Human Coiled-coil domain-containing protein 24 protein (1-307AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction