Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113163
Description
CCDC26 Polyclonal Antibody for Western Blot, ELISA
CCDC26 is a protein coding gene.
Specifications
| CCDC26 | |
| Polyclonal | |
| Unconjugated | |
| CCDC26 | |
| CCDC26; CCDC26 long non-coding RNA; RAM | |
| Recombinant Human Putative coiled-coil domain-containing protein 26 protein (1-109AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8TAB7 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 137196 | |
| -20°C or -80°C if preferred | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction