Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543943
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543943 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543943 Supplier Invitrogen™ Supplier No. PA543943
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RALVLLQSMA SRDARCPEWK PHQKPRTLSK SVQTISRYYR KTSEPKDAAS Sequence homology: Human: 100%.

The gene CCDC27 is part of a family of genes that encode coiled-coil domain-containing proteins. These proteins are characterized by their structural motif-a coiled-coil domain, which consists of two or more alpha helices coiled together. While specific details about CCDC27 are not thoroughly documented in the literature, coiled-coil domain proteins are known for their diverse roles in cellular structure and function, including participation in intracellular transport, signal transduction, and cellular scaffolding. Although the role of CCDC27 within these processes remains speculative, similar coiled-coil domain proteins are implicated in cancer pathogenesis due to their involvement in cellular processes that are frequently dysregulated in oncogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC27
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCDC27
Gene Accession No. Q2M243
Gene Alias CCDC27; coiled-coil domain containing 27; coiled-coil domain-containing protein 27; RP1-286D6.1
Gene Symbols CCDC27
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CCDC27 (aa 65-114).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148870
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.