Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC28A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563069
Description
Immunogen sequence: LARLNLELYG ELEELPEDKR KTASDSNLDR LLSDLEELNS SIQKLHLADA QDVPNTSA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 88%.
This gene encodes a coiled-coil domain containing protein. Although the specific function of this gene has not yet been determined, this gene is a known translocation partner of nucleoporin 98 in acute leukemias. The resulting fusion gene produces a nucleoporin 98-coiled-coil domain-containing protein 28A chimeric protein which may be involved in promoting myeloproliferative neoplasms.
Specifications
CCDC28A | |
Polyclonal | |
Unconjugated | |
CCDC28A | |
1700009P13Rik; AI480677; C6orf80; CCDC28A; CCRL1AP; chemokine C-C motif receptor-like 1 adjacent; coiled-coil domain containing 28A; coiled-coil domain-containing protein 28A; RGD1310326 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25901 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IWP9 | |
CCDC28A | |
Recombinant protein corresponding to Human CCDC28A. Recombinant protein control fragment (Product #RP-101716). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction