Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC28B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555909 Supplier Invitrogen™ Supplier No. PA555909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEDLSNSMYP FQGTRLCVCV PERSVSSSPA LQEYSHTTNF PTSCSPVRFS HRLPKPRYRN LEFP Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 29%.

The product of this gene localizes to centrosomes and basal bodies. It interacts and colocalizes with several proteins associated with Bardet-Biedl syndrome (BBS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC28B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC28B
Gene Accession No. Q9BUN5
Gene Alias 1110002E08Rik; 1810010N17Rik; Ccdc28b; coiled coil domain containing 28B; coiled-coil domain containing 28B; coiled-coil domain-containing protein 28B; RP4-622L5.5
Gene Symbols CCDC28B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC28B. Recombinant protein control fragment (Product #RP-94345).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79140
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.