Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537980
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537980 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537980 Supplier Invitrogen™ Supplier No. PA537980
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse, rat, dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

Negatively regulates TNF-alpha-induced pro-inflammatory response in endothelial cells (ECs) via inhibition of TNF-alpha-induced NF-kappaB activation in ECs (PubMed:25193116). Positively regulates lipid accumulation in adipose cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CCDC3
Gene Accession No. Q9BQI4
Gene Alias 2310045O21Rik; Ccdc3; coiled-coil domain containing 3; coiled-coil domain-containing protein 3; Fat/vessel-derived secretory protein; favine
Gene Symbols CCDC3
Host Species Goat
Immunogen Peptide with sequence C-PHGVNFQDAIFPDTQ, from the internal region of CCDC3 (aa134-148).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83643
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.