Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC3 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. PIPA5122149
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122149 100 μL
PIPA5122150 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122149 Supplier Invitrogen™ Supplier No. PA5122149
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Negatively regulates TNF-alpha-induced pro-inflammatory response in endothelial cells (ECs) via inhibition of TNF-alpha-induced NF-kappaB activation in ECs (PubMed:25193116). Positively regulates lipid accumulation in adipose cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CCDC3
Gene Accession No. Q9BQI4, Q9D6Y1
Gene Alias 2310045O21Rik; Ccdc3; coiled-coil domain containing 3; coiled-coil domain-containing protein 3; Fat/vessel-derived secretory protein; favine
Gene Symbols CCDC3
Host Species Rabbit
Immunogen Cys22-Ala251.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498795, 74186, 83643
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.